Product Details

SNP ID
rs4930409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67070494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCGTGGGCTGCAAGACTGACCTG[C/T]GCAAGGACAAATCACTGGTGAACAA
Phenotype
MIM: 605781
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHOD PubMed Links

Gene Details

Gene
RHOD
Gene Name
ras homolog family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300886.1 485 Missense Mutation CGC,TGC R68C NP_001287815.1
NM_014578.3 485 Missense Mutation CGC,TGC R134C NP_055393.1

View Full Product Details