Product Details

SNP ID
rs12422858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122924208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCAGGGAGTGTGGGGACCTG[A/G]GTTCCAGCCTTGGCCATTCTGCTAA
Phenotype
MIM: 605453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCB9 PubMed Links

Gene Details

Gene
ABCB9
Gene Name
ATP binding cassette subfamily B member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243013.1 Intron NP_001229942.1
NM_001243014.1 Intron NP_001229943.1
NM_019624.3 Intron NP_062570.1
NM_019625.3 Intron NP_062571.1
NM_203444.3 Intron NP_982269.2
XM_005253558.2 Intron XP_005253615.1
XM_011538095.2 Intron XP_011536397.1
XM_011538096.2 Intron XP_011536398.1
XM_011538098.2 Intron XP_011536400.1
XM_011538099.2 Intron XP_011536401.1
XM_017019103.1 Intron XP_016874592.1
XM_017019104.1 Intron XP_016874593.1

View Full Product Details