Product Details

SNP ID
rs10877019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57779146 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAATTACATTCAGGCTGCCAAGA[C/T]TTTGTCCTCTATGCCCTGGGGTCTC
Phenotype
MIM: 604466 MIM: 615258 MIM: 604723
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METTL1 PubMed Links

Gene Details

Gene
METTL1
Gene Name
methyltransferase like 1
There are no transcripts associated with this gene.

Gene
METTL21B
Gene Name
methyltransferase like 21B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015433.2 Intron NP_056248.2
NM_206914.1 Intron NP_996797.1
Gene
TSFM
Gene Name
Ts translation elongation factor, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details