Product Details

SNP ID
rs13418570
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:1416256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCTTCCCAGAATAAATCAGTCAC[A/T]CTATCACCTGGAACATGCTAGATGT
Phenotype
MIM: 606765
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
TPO PubMed Links

Gene Details

Gene
TPO
Gene Name
thyroid peroxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000547.5 Intron NP_000538.3
NM_001206744.1 Intron NP_001193673.1
NM_001206745.1 Intron NP_001193674.1
NM_175719.3 Intron NP_783650.1
NM_175721.3 Intron NP_783652.1
NM_175722.3 Intron NP_783653.1
XM_011510379.2 Intron XP_011508681.1
XM_011510380.2 Intron XP_011508682.1
XM_011510381.2 Intron XP_011508683.1

View Full Product Details