Product Details

SNP ID
rs12608868
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35283194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGTGGAGTCTTATTCTAGACAG[C/T]GGGGACGAGGCCATGAACAAGGTAG
Phenotype
MIM: 606464 MIM: 159460 MIM: 600390
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HAMP PubMed Links
Additional Information
For this assay, SNP(s) [rs111623180] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HAMP
Gene Name
hepcidin antimicrobial peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021175.3 Intron NP_066998.1
Gene
MAG
Gene Name
myelin associated glycoprotein
There are no transcripts associated with this gene.

Gene
USF2
Gene Name
upstream transcription factor 2, c-fos interacting
There are no transcripts associated with this gene.

View Full Product Details