Product Details

SNP ID
rs12489552
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46705800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGACCCACAGCCCAGCACGG[C/G]CCCACCCAAGCCCAAGCCGCCTCCG
Phenotype
MIM: 607950 MIM: 607237
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PRSS50 PubMed Links
Additional Information
For this assay, SNP(s) [rs74548422] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PRSS50
Gene Name
protease, serine 50
There are no transcripts associated with this gene.

Gene
TMIE
Gene Name
transmembrane inner ear
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147196.2 830 Missense Mutation GCC,GGC A35G NP_671729.2
XM_006713097.3 830 UTR 5 XP_006713160.1

View Full Product Details