Product Details

SNP ID
rs11152546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2788903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCTCATGGTAAACTTGAGGAAC[A/G]TGCAAAAGTCATTGCAAAAAGTTTT
Phenotype
MIM: 300879
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XG PubMed Links

Gene Details

Gene
XG
Gene Name
Xg blood group
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141919.1 Intron NP_001135391.1
NM_001141920.1 Intron NP_001135392.1
NM_175569.2 Intron NP_780778.1
XM_005274587.3 Intron XP_005274644.1
XM_011545575.2 Intron XP_011543877.1
XM_017029787.1 Intron XP_016885276.1

View Full Product Details