Product Details

SNP ID
rs3772071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:160279033 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTGCCTGCCTAGGGTTACAAAA[C/T]GCTTTCTTAAAAAAAAATCTGGAAA
Phenotype
MIM: 602310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100505984 PubMed Links

Gene Details

Gene
LOC100505984
Gene Name
uncharacterized LOC100505984
There are no transcripts associated with this gene.

Gene
RBMS1
Gene Name
RNA binding motif single stranded interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002897.4 Intron NP_002888.1
NM_016836.3 Intron NP_058520.1
XM_005246737.1 Intron XP_005246794.1
XM_005246738.2 Intron XP_005246795.1
XM_005246739.2 Intron XP_005246796.1
XM_005246740.1 Intron XP_005246797.1
XM_005246741.3 Intron XP_005246798.1
XM_017004626.1 Intron XP_016860115.1

View Full Product Details