Product Details
- SNP ID
-
rs11652712
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:19337659 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCCAGGTCTCAGCGGCTTCCCGC[A/G]GAAGTTTCTCACAGAAGTCAGTGAC
- Phenotype
-
MIM: 614144
MIM: 607263
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
B9D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78797942] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- B9D1
- Gene Name
- B9 domain containing 1
- Gene
- EPN2
- Gene Name
- epsin 2
- Gene
- MIR1180
- Gene Name
- microRNA 1180
There are no transcripts associated with this gene.
View Full Product Details