Product Details
- SNP ID
-
rs12077882
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:154330417 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGAAGAAGAAAGGAGGGCGCGGG[C/G]TAATGACCGAGAATACAATGAGAAA
- Phenotype
-
MIM: 606578
MIM: 605867
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AQP10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11430764] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AQP10
- Gene Name
- aquaporin 10
There are no transcripts associated with this gene.
- Gene
- ATP8B2
- Gene Name
- ATPase phospholipid transporting 8B2
View Full Product Details