Product Details

SNP ID
rs12063890
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:154164115 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTATCTTCTTTCCCTTCCCCCAG[A/C]TCCCCAAACCAGGACAACATGATCT
Phenotype
MIM: 191030
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NUP210L PubMed Links

Gene Details

Gene
NUP210L
Gene Name
nucleoporin 210 like
There are no transcripts associated with this gene.

Gene
TPM3
Gene Name
tropomyosin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043351.1 4797 Intron NP_001036816.1
NM_001043352.1 4797 Intron NP_001036817.1
NM_001043353.1 4797 Intron NP_001036818.1
NM_001278188.1 4797 Intron NP_001265117.1
NM_001278189.1 4797 Intron NP_001265118.1
NM_001278190.1 4797 Intron NP_001265119.1
NM_001278191.1 4797 Intron NP_001265120.1
NM_152263.3 4797 UTR 3 NP_689476.2
NM_153649.3 4797 Intron NP_705935.1

View Full Product Details