Product Details

SNP ID
rs7895458
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46290653 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTGTGAAGTTGTATCTCCTTGA[C/G]GCTTTCATTGGCATTTCCCTAGTGA
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ANTXRL PubMed Links

Gene Details

Gene
ANTXRL
Gene Name
anthrax toxin receptor-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278688.2 Intron NP_001265617.1
XM_006717684.3 Intron XP_006717747.1
XM_011539430.2 Intron XP_011537732.1
XM_011539431.2 Intron XP_011537733.1
XM_011539433.2 Intron XP_011537735.1
XM_011539434.2 Intron XP_011537736.1
XM_011539437.2 Intron XP_011537739.1
XM_011539438.2 Intron XP_011537740.1
XM_011539439.2 Intron XP_011537741.1
XM_011539440.2 Intron XP_011537742.1
XM_011539441.1 Intron XP_011537743.1
XM_011539442.1 Intron XP_011537744.1
XM_011539443.1 Intron XP_011537745.1
XM_017015818.1 Intron XP_016871307.1
XM_017015819.1 Intron XP_016871308.1
Gene
LOC105378577
Gene Name
basic proline-rich protein-like
There are no transcripts associated with this gene.

View Full Product Details