Product Details

SNP ID
rs11592871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75285903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCATGTTCTAGACCTGGCTTTGC[A/G]GCTGAGGGAGCGGGTCTTGAGTTGT
Phenotype
MIM: 613902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF503 PubMed Links

Gene Details

Gene
ZNF503
Gene Name
zinc finger protein 503
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032772.5 Intron NP_116161.2

View Full Product Details