Product Details

SNP ID
rs11001360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75286653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGCCTGACTGAGATGCACATCA[G/T]ATGCTGGGAAGAGACTTGGTGCATG
Phenotype
MIM: 613902
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF503 PubMed Links

Gene Details

Gene
ZNF503
Gene Name
zinc finger protein 503
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032772.5 Intron NP_116161.2
Gene
ZNF503-AS1
Gene Name
ZNF503 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details