Product Details

SNP ID
rs12195971
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26460196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGTCTTGCTCTGTTGTACCCTC[A/G]CTGGCACGATCTTGTCTCACTGCAA
Phenotype
MIM: 613590 MIM: 613594 MIM: 613595
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BTN2A1 PubMed Links
Additional Information
For this assay, SNP(s) [rs141410781] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
BTN2A1
Gene Name
butyrophilin subfamily 2 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197233.2 Intron NP_001184162.1
NM_001197234.2 Intron NP_001184163.1
NM_007049.4 Intron NP_008980.1
NM_078476.3 Intron NP_510961.1
Gene
BTN3A2
Gene Name
butyrophilin subfamily 3 member A2
There are no transcripts associated with this gene.

Gene
BTN3A3
Gene Name
butyrophilin subfamily 3 member A3
There are no transcripts associated with this gene.

View Full Product Details