Product Details

SNP ID
rs12239030
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:108891035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATGCTTTCCGGAATGTACTTCA[A/G]ACCAATCTGGGGTTGGAAGAGTAAC
Phenotype
MIM: 609245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPSM2 PubMed Links

Gene Details

Gene
GPSM2
Gene Name
G-protein signaling modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321038.1 Intron NP_001307967.1
NM_001321039.1 Intron NP_001307968.1
NM_013296.4 Intron NP_037428.3
XM_006710589.2 Intron XP_006710652.1
XM_011541301.2 Intron XP_011539603.1
XM_011541302.2 Intron XP_011539604.1
XM_011541303.2 Intron XP_011539605.1
XM_017001097.1 Intron XP_016856586.1
XM_017001098.1 Intron XP_016856587.1

View Full Product Details