Product Details

SNP ID
rs12144309
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:113772871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAATCTCTTCCTCTATACTGAAG[C/T]CAATTCCAGGTGGATTAAATATGTA
Phenotype
MIM: 615858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSBN1 PubMed Links

Gene Details

Gene
RSBN1
Gene Name
round spermatid basic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018364.4 Intron NP_060834.2
XM_017001518.1 Intron XP_016857007.1

View Full Product Details