Product Details

SNP ID
rs12062220
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:114397718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGGCTGTATAAAGTCTTCATCA[A/G]AGTCCTCAGTTACCTCACCATCATC
Phenotype
MIM: 605769
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM33 PubMed Links

Gene Details

Gene
TRIM33
Gene Name
tripartite motif containing 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015906.3 2673 Missense Mutation TCT,TTT S1105F NP_056990.3
NM_033020.2 2673 Missense Mutation TCT,TTT S1088F NP_148980.2
XM_005270936.3 2673 Missense Mutation TCT,TTT S1129F XP_005270993.1
XM_005270937.3 2673 Missense Mutation TCT,TTT S1112F XP_005270994.1
XM_011541568.2 2673 Missense Mutation TCT,TTT S1128F XP_011539870.1
XM_017001452.1 2673 Missense Mutation TCT,TTT S713F XP_016856941.1
XM_017001453.1 2673 Missense Mutation TCT,TTT S696F XP_016856942.1
XM_017001454.1 2673 Intron XP_016856943.1

View Full Product Details