Product Details

SNP ID
rs12302991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50339692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATCTTTCCAGTCTCATCTCTTA[C/G]TACATTTTGCTTTTTCTTTGTGTTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM186A PubMed Links

Gene Details

Gene
FAM186A
Gene Name
family with sequence similarity 186 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145475.1 Intron NP_001138947.1
XM_006719231.3 Intron XP_006719294.1
XM_011537890.2 Intron XP_011536192.1
XM_011537892.1 Intron XP_011536194.1

View Full Product Details