Product Details

SNP ID
rs11837677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50924678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTTCTTTTAGAGGTCACTGGA[A/G]AGCCCGGTGGGGTGTAAGTACACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL7A PubMed Links

Gene Details

Gene
METTL7A
Gene Name
methyltransferase like 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014033.3 282 Intron NP_054752.3
XM_006719332.1 282 UTR 5 XP_006719395.1

View Full Product Details