Product Details

SNP ID
rs11169588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50845487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCACCATGTGCTGTCAACTCCTA[G/T]GTCTCTCCTGATGAGTCATATTTCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS12 PubMed Links

Gene Details

Gene
TMPRSS12
Gene Name
transmembrane protease, serine 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182559.2 Intron NP_872365.1

View Full Product Details