Product Details

SNP ID
rs4645741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35401776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGAGTGAGTCGTGAAGACCAAC[A/G]GATAACTTCCACGGTGGTCATTTCC
Phenotype
MIM: 141250 MIM: 602696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMOX1 PubMed Links

Gene Details

Gene
HMOX1
Gene Name
heme oxygenase 1
There are no transcripts associated with this gene.

Gene
MCM5
Gene Name
minichromosome maintenance complex component 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006739.3 Intron NP_006730.2
XM_006724242.3 Intron XP_006724305.1

View Full Product Details