Product Details

SNP ID
rs7292295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36967361 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGAGAAATGGAGGAGTTTCCAG[A/G]CCCACGTGACACTGAATCACAATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LL22NC01-81G9.3 PubMed Links

Gene Details

Gene
LL22NC01-81G9.3
Gene Name
uncharacterized protein FLJ39582-like
There are no transcripts associated with this gene.

View Full Product Details