Product Details

SNP ID
rs4264986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140613681 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGTTTTCCGTCTACTGTTTCTA[A/T]CTTCATACAATCATGCTATTTCTAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMCO6 PubMed Links

Gene Details

Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 Intron NP_001287909.1
NM_001300982.1 Intron NP_001287911.1
NM_018502.4 Intron NP_060972.3
XM_005268477.1 Intron XP_005268534.1
XM_011537663.2 Intron XP_011535965.1
XM_011537665.2 Intron XP_011535967.1
XM_011537668.2 Intron XP_011535970.1
XM_017009617.1 Intron XP_016865106.1
XM_017009618.1 Intron XP_016865107.1
XM_017009619.1 Intron XP_016865108.1
XM_017009620.1 Intron XP_016865109.1

View Full Product Details