Product Details

SNP ID
rs11956826
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141095114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGAGCCCTGTGTCCTACCTTT[C/G]CAGGTGTTACTAGAAAATCCCTTGC
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018936.3 508 Missense Mutation TTC,TTG F108L NP_061759.1
Gene
PCDHB3
Gene Name
protocadherin beta 3
There are no transcripts associated with this gene.

View Full Product Details