Product Details

SNP ID
rs11792542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92221390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCCAATAAATCCCAGGCTATCA[C/G]GTGTTACAAATAAAAGGATGGGATG
Phenotype
MIM: 600709
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IARS PubMed Links

Gene Details

Gene
IARS
Gene Name
isoleucyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002161.5 Intron NP_002152.2
NM_013417.3 Intron NP_038203.2

View Full Product Details