Product Details

SNP ID
rs4271002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:18390758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACAAAGCCATATGATACATGT[C/G]ACACTTATACCACATGCATAGAGTT
Phenotype
MIM: 612182
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NAT2 PubMed Links

Gene Details

Gene
NAT2
Gene Name
N-acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000015.2 Intron NP_000006.2
XM_017012938.1 Intron XP_016868427.1

View Full Product Details