Product Details

SNP ID
rs11716582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47592269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGTCTTTGCCAGCCATATGGTC[C/T]GTGTCACAACTATTCAACTCTGCAA
Phenotype
MIM: 601732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMARCC1 PubMed Links

Gene Details

Gene
SMARCC1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003074.3 Intron NP_003065.3
XM_011534034.1 Intron XP_011532336.1

View Full Product Details