Product Details

SNP ID
rs11659867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74433649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATTGCCAGTTGACATTGGCCA[A/T]CTGACTCATTGACCAGTCTAGGTAT
Phenotype
MIM: 614544
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM69C PubMed Links

Gene Details

Gene
FAM69C
Gene Name
family with sequence similarity 69 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044369.2 4167 Intron NP_001037834.2
XM_011525816.2 4167 UTR 3 XP_011524118.1
XM_017025551.1 4167 UTR 3 XP_016881040.1

View Full Product Details