Product Details
- SNP ID
-
rs11212749
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:108668877 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGAGGATGGTGTAATTAGGGGAA[A/G]TTGAGTTATCCTATACTGAGCTCCT
- Phenotype
-
MIM: 601235
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114211399,rs115740831] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX10
- Gene Name
- DEAD-box helicase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004398.3 |
|
Intron |
|
|
NP_004389.2 |
View Full Product Details