Product Details
- SNP ID
-
rs12226431
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47784304 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGATGCAGACAGTCTAAGAACCC[A/G]TTTTTGATTTTTAATAGGTGAAACA
- Phenotype
-
MIM: 607614
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP160
PubMed Links
Gene Details
- Gene
- NUP160
- Gene Name
- nucleoporin 160
View Full Product Details