Product Details

SNP ID
rs12577282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47611809 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTCTTTTTCAGAGTTGCAATAA[A/G]ATGATTCCAGAGCAATTACAGTTTT
Phenotype
MIM: 613221
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTCH2 PubMed Links

Gene Details

Gene
MTCH2
Gene Name
mitochondrial carrier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317231.1 Intron NP_001304160.1
NM_001317232.1 Intron NP_001304161.1
NM_001317233.1 Intron NP_001304162.1
NM_014342.3 Intron NP_055157.1
XM_006718172.2 Intron XP_006718235.1
XM_011519959.2 Intron XP_011518261.1
XM_011519960.2 Intron XP_011518262.1
XM_011519961.2 Intron XP_011518263.1
XM_017017462.1 Intron XP_016872951.1
XM_017017463.1 Intron XP_016872952.1

View Full Product Details