Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001258419.1 | 7682 | Intron | NP_001245348.1 | ||
NM_020929.2 | 7682 | Intron | NP_065980.1 | ||
XM_011520238.2 | 7682 | UTR 3 | XP_011518540.1 | ||
XM_011520239.2 | 7682 | UTR 3 | XP_011518541.1 | ||
XM_011520240.2 | 7682 | UTR 3 | XP_011518542.1 | ||
XM_011520241.2 | 7682 | UTR 3 | XP_011518543.1 | ||
XM_011520242.2 | 7682 | UTR 3 | XP_011518544.1 | ||
XM_011520243.2 | 7682 | UTR 3 | XP_011518545.1 | ||
XM_011520244.2 | 7682 | UTR 3 | XP_011518546.1 | ||
XM_017018070.1 | 7682 | UTR 3 | XP_016873559.1 | ||
XM_017018071.1 | 7682 | UTR 3 | XP_016873560.1 | ||
XM_017018072.1 | 7682 | UTR 3 | XP_016873561.1 | ||
XM_017018073.1 | 7682 | UTR 3 | XP_016873562.1 | ||
XM_017018074.1 | 7682 | UTR 3 | XP_016873563.1 | ||
XM_017018075.1 | 7682 | UTR 3 | XP_016873564.1 | ||
XM_017018076.1 | 7682 | UTR 3 | XP_016873565.1 | ||
XM_017018077.1 | 7682 | UTR 3 | XP_016873566.1 | ||
XM_017018078.1 | 7682 | UTR 3 | XP_016873567.1 | ||
XM_017018079.1 | 7682 | UTR 3 | XP_016873568.1 |