Product Details

SNP ID
rs11033706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36600549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTGAAACTGTCCCCCTCACTC[C/T]GGTCCATGGAAAAATTGTCCTCCAC
Phenotype
MIM: 179616
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf74 PubMed Links

Gene Details

Gene
C11orf74
Gene Name
chromosome 11 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276722.1 Intron NP_001263651.1
NM_001276723.1 Intron NP_001263652.1
NM_001276724.1 Intron NP_001263653.1
NM_001276725.1 Intron NP_001263654.1
NM_001276726.1 Intron NP_001263655.1
NM_001276727.1 Intron NP_001263656.1
NM_138787.3 Intron NP_620142.2
XM_006718138.3 Intron XP_006718201.1
XM_011519885.2 Intron XP_011518187.1
Gene
RAG2
Gene Name
recombination activating gene 2
There are no transcripts associated with this gene.

View Full Product Details