Product Details

SNP ID
rs10836528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36297111 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTATGGATGAGGAAATTGAAGT[A/G]CGGATAATTTAAGTCATTCACCCGG
Phenotype
MIM: 612299 MIM: 611728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMMD9 PubMed Links

Gene Details

Gene
COMMD9
Gene Name
COMM domain containing 9
There are no transcripts associated with this gene.

Gene
PRR5L
Gene Name
proline rich 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160167.1 Intron NP_001153639.1
NM_001160168.1 Intron NP_001153640.1
NM_001160169.1 Intron NP_001153641.1
NM_024841.4 Intron NP_079117.3

View Full Product Details