Product Details

SNP ID
rs12282742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18244252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTTGGCAAGATACAGTTCACT[C/T]CAGAGCACTGCCACTTCCACAGCTG
Phenotype
MIM: 104751 MIM: 104752
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAA2 PubMed Links

Gene Details

Gene
SAA2
Gene Name
serum amyloid A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127380.2 Intron NP_001120852.1
NM_030754.4 Intron NP_110381.2
Gene
SAA2-SAA4
Gene Name
SAA2-SAA4 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199744.1 Intron NP_001186673.1
Gene
SAA4
Gene Name
serum amyloid A4, constitutive
There are no transcripts associated with this gene.

View Full Product Details