Product Details

SNP ID
rs11821600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18269906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCTGGCTGTGAGGCCCTCAGGG[C/T]AGGGATACAAAGCGGGGAGAGGGTA
Phenotype
MIM: 607521 MIM: 104750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPS5 PubMed Links

Gene Details

Gene
HPS5
Gene Name
HPS5, biogenesis of lysosomal organelles complex 2 subunit 2
There are no transcripts associated with this gene.

Gene
SAA1
Gene Name
serum amyloid A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000331.5 557 UTR 3 NP_000322.2
NM_001178006.2 557 UTR 3 NP_001171477.1
NM_199161.4 557 UTR 3 NP_954630.1
Gene
ST13P5
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details