Product Details

SNP ID
rs10840110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8682544 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGGGCGGCGTGCAGTACAGCACA[C/T]CTAGCCCCGATTCTTCAACAGTTCT
Phenotype
MIM: 603637 MIM: 611333 MIM: 612000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL27A PubMed Links

Gene Details

Gene
RPL27A
Gene Name
ribosomal protein L27a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000990.4 97 UTR 5 NP_000981.1
Gene
SNORA3A
Gene Name
small nucleolar RNA, H/ACA box 3A
There are no transcripts associated with this gene.

Gene
SNORA3B
Gene Name
small nucleolar RNA, H/ACA box 3B
There are no transcripts associated with this gene.

Gene
TRIM66
Gene Name
tripartite motif containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014818.1 97 Intron NP_055633.1
XM_006718397.2 97 Intron XP_006718460.1
XM_006718398.3 97 Intron XP_006718461.1
XM_011520504.1 97 Intron XP_011518806.1
XM_011520507.2 97 Intron XP_011518809.1
XM_011520508.1 97 Intron XP_011518810.1
XM_011520509.1 97 Intron XP_011518811.1
XM_011520510.1 97 Intron XP_011518812.1
XM_011520511.1 97 Intron XP_011518813.1
XM_011520512.1 97 Intron XP_011518814.1
XM_011520513.1 97 Intron XP_011518815.1
XM_011520514.2 97 Intron XP_011518816.1
XM_011520515.1 97 Intron XP_011518817.1
XM_011520516.1 97 Intron XP_011518818.1
XM_011520517.2 97 Intron XP_011518819.1
XM_011520518.1 97 Intron XP_011518820.1
XM_011520519.1 97 Intron XP_011518821.1
XM_011520522.2 97 Intron XP_011518824.1
XM_011520523.2 97 Intron XP_011518825.1
XM_011520524.1 97 Intron XP_011518826.1
XM_011520525.1 97 Intron XP_011518827.1
XM_011520526.2 97 Intron XP_011518828.1
XM_011520527.1 97 Intron XP_011518829.1
XM_017018629.1 97 Intron XP_016874118.1
XM_017018630.1 97 Intron XP_016874119.1
XM_017018631.1 97 Intron XP_016874120.1

View Full Product Details