Product Details

SNP ID
hCV31044119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7027148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTCCAGTAATTCCCCTCCCTTT[A/C]AATGTCAATTTTTCCCTCTATACTA
Phenotype
MIM: 609665 MIM: 605015
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NLRP14 PubMed Links

Gene Details

Gene
NLRP14
Gene Name
NLR family pyrin domain containing 14
There are no transcripts associated with this gene.

Gene
ZNF214
Gene Name
zinc finger protein 214
There are no transcripts associated with this gene.

View Full Product Details