Product Details

SNP ID
rs12287453
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7025707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATGAGCAAAAATGTCAAATAAT[C/T]GAAGGTTAATTTAAGAACTAAAGAA
Phenotype
MIM: 609665 MIM: 605015
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NLRP14 PubMed Links

Gene Details

Gene
NLRP14
Gene Name
NLR family pyrin domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176822.3 Intron NP_789792.1
XM_011520044.1 Intron XP_011518346.1
Gene
ZNF214
Gene Name
zinc finger protein 214
There are no transcripts associated with this gene.

View Full Product Details