Product Details
- SNP ID
-
rs11260895
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:16709105 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATCTGAGATTCAAAGCTAGTCAAA[A/C]ATCCCGGGTTTTGTTGTTGCTGTTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ESPNP
PubMed Links
Gene Details
- Gene
- ESPNP
- Gene Name
- espin pseudogene
There are no transcripts associated with this gene.
View Full Product Details