Product Details

SNP ID
rs11164006
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:84556184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTATCTAGTACTAACACAGTCT[C/G]TCTCAGGCAGCACATGAGCTATTGG
Phenotype
MIM: 600873
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTBS PubMed Links

Gene Details

Gene
CTBS
Gene Name
chitobiase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004388.2 1844 Intron NP_004379.1
Gene
SPATA1
Gene Name
spermatogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310156.1 1844 UTR 3 NP_001297085.1
XM_011542514.2 1844 Intron XP_011540816.1
XM_011542515.2 1844 Intron XP_011540817.1
XM_011542516.1 1844 Intron XP_011540818.1
XM_011542517.1 1844 Intron XP_011540819.1
XM_011542519.2 1844 Intron XP_011540821.1
XM_011542520.2 1844 Intron XP_011540822.1
XM_011542521.2 1844 Intron XP_011540823.1
XM_011542522.2 1844 Intron XP_011540824.1

View Full Product Details