Product Details

SNP ID
rs12158549
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46299671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCGGCTTCTACTTCCCGTTGG[A/C]TAATATGTGCATTTGTAATGTCAGG
Phenotype
MIM: 607477
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GTSE1 PubMed Links

Gene Details

Gene
GTSE1
Gene Name
G2 and S-phase expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016426.6 Intron NP_057510.4
XM_005261627.3 Intron XP_005261684.1
XM_011530211.2 Intron XP_011528513.1
XM_011530213.2 Intron XP_011528515.1
XM_017028815.1 Intron XP_016884304.1
XM_017028816.1 Intron XP_016884305.1
Gene
GTSE1-AS1
Gene Name
GTSE1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
There are no transcripts associated with this gene.

View Full Product Details