Product Details

SNP ID
rs7270791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44496488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACAACTCATTATGTGAATACTC[A/G]CTTTAGCCTGAATGTGTAAAAACAG
Phenotype
MIM: 607165
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERINC3 PubMed Links

Gene Details

Gene
SERINC3
Gene Name
serine incorporator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006811.3 Intron NP_006802.1
NM_198941.2 Intron NP_945179.1
XM_017027601.1 Intron XP_016883090.1
Gene
TTPAL
Gene Name
alpha tocopherol transfer protein like
There are no transcripts associated with this gene.

View Full Product Details