Product Details

SNP ID
rs12025728
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:247035508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTAGGCAACTGAAATATGTTAGG[C/T]TACAGACTGATAAGATCAAAATAGT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF670 PubMed Links

Gene Details

Gene
ZNF670
Gene Name
zinc finger protein 670
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204220.1 3328 UTR 3 NP_001191149.1
NM_033213.4 3328 UTR 3 NP_149990.1
Gene
ZNF670-ZNF695
Gene Name
ZNF670-ZNF695 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details