Product Details

SNP ID
rs13224456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64666718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCCATCTTTTTTCTCTTAAAAA[G/T]CTATGGAAGTCACCACAAAAATGTT
Phenotype
MIM: 603989
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF107 PubMed Links

Gene Details

Gene
ZNF107
Gene Name
zinc finger protein 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013746.2 Intron NP_001013768.1
NM_001282359.1 Intron NP_001269288.1
NM_001282360.1 Intron NP_001269289.1
NM_016220.4 Intron NP_057304.1
XM_017012283.1 Intron XP_016867772.1
XM_017012284.1 Intron XP_016867773.1
XM_017012285.1 Intron XP_016867774.1
XM_017012286.1 Intron XP_016867775.1

View Full Product Details