Product Details

SNP ID
rs7151963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96513597 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGATGTAACAAGTTTGCTTTCAT[C/T]GTGATGAATAAGGGCTCGTTTTTTT
Phenotype
MIM: 605553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAPOLA PubMed Links

Gene Details

Gene
PAPOLA
Gene Name
poly(A) polymerase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252006.1 Intron NP_001238935.1
NM_001252007.1 Intron NP_001238936.1
NM_001293627.1 Intron NP_001280556.1
NM_001293628.1 Intron NP_001280557.1
NM_001293632.1 Intron NP_001280561.1
NM_032632.4 Intron NP_116021.2
XM_005267281.1 Intron XP_005267338.1
XM_005267282.2 Intron XP_005267339.1
XM_017020935.1 Intron XP_016876424.1
XM_017020936.1 Intron XP_016876425.1

View Full Product Details