Product Details

SNP ID
rs12884005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100881071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCACACTGACGGTAACGTTGCA[A/G]GTCGTCTTGCAGGGCTTCTCGCAAG
Phenotype
MIM: 611709 MIM: 611710 MIM: 611708 MIM: 611711 MIM: 611896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR127 PubMed Links

Gene Details

Gene
MIR127
Gene Name
microRNA 127
There are no transcripts associated with this gene.

Gene
MIR136
Gene Name
microRNA 136
There are no transcripts associated with this gene.

Gene
MIR337
Gene Name
microRNA 337
There are no transcripts associated with this gene.

Gene
MIR431
Gene Name
microRNA 431
There are no transcripts associated with this gene.

Gene
MIR432
Gene Name
microRNA 432
There are no transcripts associated with this gene.

Gene
MIR433
Gene Name
microRNA 433
There are no transcripts associated with this gene.

Gene
MIR665
Gene Name
microRNA 665
There are no transcripts associated with this gene.

Gene
RTL1
Gene Name
retrotransposon-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134888.2 3777 Silent Mutation CTG,TTG L1240L NP_001128360.1

View Full Product Details