Product Details

SNP ID
rs12149002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55732064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTTTGCTTCCAATCTGCATGA[G/T]CACCCTATCTGCCCCTGAGCTTAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CES1P2 PubMed Links

Gene Details

Gene
CES1P2
Gene Name
carboxylesterase 1 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details