Product Details
- SNP ID
-
rs6969815
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:138801791 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATTTCTTCTTCTTCTATGCACGTT[C/T]TTCCTCTTCTTGCCTACTTCTAGCC
- Phenotype
-
MIM: 605239
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP6V0A4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112555715] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP6V0A4
- Gene Name
- ATPase H+ transporting V0 subunit a4
There are no transcripts associated with this gene.
- Gene
- TMEM213
- Gene Name
- transmembrane protein 213
View Full Product Details